Learn More
Sino Biological Recombinant Human 4-1BB/CD137 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10041H08HB20
Encompass_Preferred
A DNA sequence encoding the Human TNFRSF9 (NP_001552.2) (Met1-Gln186) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-161-3860