Learn More
Sino Biological Anti-Langerin/CD207 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100414T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human cd207, and purified by antigen affinity chromatography.
Catalog No. 50-161-1108