Learn More
Sino Biological Recombinant Human 2B4/CD244 Protein (hFc Tag)
Supplier:  Sino Biological 10042H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg221) of human 2B4 / CD244 (NP_057466.1) precursor was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-3861