Learn More
Sino Biological Anti-Lck Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10043T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Lck (rh Lck; Catalog#10043-H09B; NP_001036236.1; Met1-Pro509). Lck specific IgG was purified by Human Lck affinity chromatography.
Catalog No. 50-161-1194