Learn More
Sino Biological Anti-NEUROD1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100436T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human NeuroD1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1185