Learn More
Sino Biological Anti-GSK-3 beta/GSK3B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10044T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GSK3B (rh GSK3B; Catalog#10044-H07B; NP_002084.2; Met1-Thr433). GSK3B specific IgG was purified by Human GSK3B affinity chromatography.
Catalog No. 50-161-1219