Learn More
Sino Biological Anti-A4GALT Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100447T32100
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the C-terminus of the human A4GALT / CD77 synthase. Human A4GALT / CD77 synthase specific IgG was purified by Human A4GALT / CD77 synthase affinity chromatography.
Catalog No. 50-161-1206