Learn More
Sino Biological Anti-CCR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100449T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CCR1/CD191, and purified by antigen affinity chromatography.
Catalog No. 50-161-1216