Learn More
Sino Biological Anti-CD166/ALCAM Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10045MM03A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD166 / ALCAM (rh CD166 / ALCAM; Catalog#10045-H08H; NP_001618.2; Met 1-Ala 526) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-1259