Learn More
Sino Biological Anti-CCR2 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 100450MM02P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the center region of the Human CCR2/CD192, and it was conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-1230