Learn More
Sino Biological Anti-CCR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100450T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CCR2/cd192, and purified by antigen affinity chromatography.
Catalog No. 50-161-1237