Learn More
Sino Biological Anti-Integrin beta 3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100458T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human Integrin beta 3/ITGB3/CD61, and purified by antigen affinity chromatography.
Catalog No. 50-161-1252