Learn More
Sino Biological Anti-SPARCL1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10046MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SPARCL1 / SPARC-like 1 (rh SPARCL1 / SPARC-like 1; Catalog#10046-H08H; NP_004675.3; Met1-Phe664). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1296