Learn More
Sino Biological Anti-SPARCL1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10046MM06F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SPARCL1 (rh SPARCL1; Catalog#10046-H08H; NP_004675.3; Met 1-Phe 664) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-1305