Learn More
Sino Biological Recombinant Anti-SPARCL1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10046R390100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARCL1 / SPARC-like 1 (rh SPARCL1 / SPARC-like 1; Catalog#10046-H08H; NP_004675.3; Met1-Phe664).
Catalog No. 50-161-1310