Learn More
Sino Biological Anti-Integrin alpha 2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100468T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human Integrin alpha 2/ITGA2/CD49B, and purified by antigen affinity chromatography.
Catalog No. 50-161-1288