Learn More
Sino Biological Recombinant Human TrkB Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10047H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-His 430) of human TrkB (NP_001007098.1) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3873