Learn More
Sino Biological Recombinant Human TrkC Protein (His & hFc Tag)
Supplier:  Sino Biological 10048H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Asp428) of human TrkC (NP_001007157.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3875