Learn More
Sino Biological Recombinant Human TrkC Protein (His Tag)
Supplier:  Sino Biological 10048H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asp 428) of human TrkC (NP_001007157.1) was expressed with a C-terminal polyhistide tag.
Catalog No. 50-161-3877