Learn More
Sino Biological Anti-ILT11/LILRA5/LIR-9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100481T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LILRA5/CD85f, and purified by antigen affinity chromatography.
Catalog No. 50-161-1349