Learn More
Sino Biological Anti-ADAM8/CD156a Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100485T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human CD156a, and purified by antigen affinity chromatography.
Catalog No. 50-161-1355