Learn More
Sino Biological Recombinant Human Caspase-7 Protein (His Tag)
Supplier:  Sino Biological 10049H08E20
Encompass_Preferred
A DNA sequence encoding the human CASP7 (P55210-1) (Met 1-Gln 303) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3880