Learn More
Sino Biological Anti-Band 3/SLC4A1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100490T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human Band 3/SLC4A1/CD233, and purified by antigen affinity chromatography.
Catalog No. 50-161-1393