Learn More
Sino Biological Anti-Caspase-3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10050MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human caspase-3 / CASP-3 (rh caspase-3 / CASP-3; Catalog#10050-H08E; Q5IS54; Ser29-His277). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1431