Learn More
Sino Biological Anti-ZMYM2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100501T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human ZMYM2, and purified by antigen affinity chromatography.
Catalog No. 50-161-1405