Learn More
Sino Biological Recombinant Anti-S100B Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100508R034100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human S100B .
Catalog No. 50-161-1426