Learn More
Sino Biological Recombinant Anti-CD13 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10051R011100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD13 / ANPEP (rh CD13 / ANPEP; Catalog#10051-H08H; NP_001141.2; Lys69-Lys967,603Met/Ile).
Catalog No. 50-161-1443