Learn More
Sino Biological Anti-PAX7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100516T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PAX7 / HUP1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1437