Learn More
Sino Biological Recombinant Human IL-12 p40/IL12B Protein (His Tag)
Supplier:  Sino Biological 10052H08H100
Encompass_Preferred
A DNA sequence encoding the p40 subunit of human IL12, termed as IL12B (NP_002178.2) (Met 1-Ser 328) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3889