Learn More
Sino Biological Anti-Myosin light chain 3/MYL3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100528T32100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human MYL3, and purified by antigen affinity chromatography.
Catalog No. 50-161-1463