Learn More
Sino Biological Recombinant Human Nucleophosmin/B23/NPM1 Protein (His Tag)
Supplier:  Sino Biological 10053H07E100
Encompass_Preferred
A DNA sequence encoding the human NPM1 isoform 1 (P06748-1) N-terminal segment (Met 9-Leu 158) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-3893