Learn More
Sino Biological Anti-Nucleophosmin/B23/NPM1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10053RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NPM1 / Nucleophosmin (rh NPM1 / Nucleophosmin; Catalog#10053-H07E; P06748-1; Met 9-Leu 158). NPM1 / Nucleophosmin specific IgG was purified by Human NPM1 / Nucleophosmin affinity chromatography.
Catalog No. 50-161-1497