Learn More
Sino Biological Anti-Nac1/NACC1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100540T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NACC1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1503