Learn More
Sino Biological Recombinant Human BAFF/BLyS/TNFSF13B Protein (hFc Tag)
Supplier:  Sino Biological 10056H01H100
Encompass_Preferred
A DNA sequence encoding the soluble form of human BAFF (Q9Y275-1) (Ala 134-Leu 285) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-3897