Learn More
Sino Biological Anti-Caspase-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100561T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human CASP3, and purified by antigen affinity chromatography.
Catalog No. 50-161-1533