Learn More
Sino Biological Anti-CD151 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100567T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human CD151, and purified by antigen affinity chromatography.
Catalog No. 50-161-1545