Learn More
Sino Biological Anti-MAP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100568T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MAP2, and purified by antigen affinity chromatography.
Catalog No. 50-161-1548