Learn More
Sino Biological Anti-IL17C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100571T32100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human IL17C, and purified by antigen affinity chromatography.
Catalog No. 50-161-1569