Learn More
Sino Biological Anti-NFIB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100574T32100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human NF1B, and purified by antigen affinity chromatography.
Catalog No. 50-161-1575