Learn More
AdipoGen 10058-F4
SDP
Catalog No. 502560959
Encompass_Preferred
Chemical. CAS 403811-55-2. Formula C12H11NOS2. MW 249.35. c-Myc is a proto-oncogene that plays an important role in cell proliferation, differentiation, and apoptosis. 5-4-Ethylphenylmethylene-2-thioxo-4-thiazolidinone is a cell-permeable thiazolidinone compound that specifically inhibits the c-Myc-Max interaction/dimerization at 64 µM, preventing c-Myc-dependent gene expression and cell proliferation. 10058-F4 inhibits tumor cell growth and proliferation in a c-Myc-dependent manner both in vitro and in vivo. 10058-F4 induces cell-cycle arrest G0/G1 phase and apoptosis. 10058-F4 has been found to also upregulate cyclin-dependent kinase CDK inhibitors, p21 and p27. In addition to c-Myc, 10058-F4 inhibits the nuclear Myc protein, N-Myc, at 50 µM, inducing arrest, apoptosis, and differentiation in neuroblastoma cells overexpressing the gene for N-Myc. This compound can be used to delineate novel actions of Myc proteins, especially those related to lipid and glucose m
Catalog No. 50-256-0959 Supplier AdipoGen Supplier No. CDXE0533M005
Chemical. CAS 403811-55-2. Formula C12H11NOS2. MW 249.35. c-Myc is a proto-oncogene that plays an important role in cell proliferation, differentiation, and apoptosis. 5-4-Ethylphenylmethylene-2-thioxo-4-thiazolidinone is a cell-permeable thiazolidinone compound that specifically inhibits the c-Myc-Max interaction/dimerization at 64 µM, preventing c-Myc-dependent gene expression and cell proliferation. 10058-F4 inhibits tumor cell growth and proliferation in a c-Myc-dependent manner both in vitro and in vivo. 10058-F4 induces cell-cycle arrest G0/G1 phase and apoptosis. 10058-F4 has been found to also upregulate cyclin-dependent kinase CDK inhibitors, p21 and p27. In addition to c-Myc, 10058-F4 inhibits the nuclear Myc protein, N-Myc, at 50 µM, inducing arrest, apoptosis, and differentiation in neuroblastoma cells overexpressing the gene for N-Myc. This compound can be used to delineate novel actions of Myc proteins, especially those related to lipid and glucose m