Learn More
Sino Biological Anti-CD157 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10060MM19P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD157/BST1 (rh CD157/BST1; Catalog#10060-H08H; NP_004325.2; Met1-Lys292) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-1660