Learn More
Sino Biological Anti-HOXB8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100606T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse HOXB8, and purified by antigen affinity chromatography.
Catalog No. 50-161-1634