Learn More
Sino Biological Anti-TLR2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10061MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TLR2 (CD282) (rh TLR2 (CD282); Catalog#10061-H08B; O60603-1; Met1-Arg587). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1685