Learn More
Sino Biological Recombinant Anti-TLR2 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10061R067A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TLR2/CD282 (rh TLR2/CD282; Catalog#10061-H08B1; NP_003255.2; Met1-Arg587) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-1691