Learn More
Sino Biological Recombinant Anti-TLR2 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10061R067P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TLR2 / CD282 (rh TLR2 / CD282; Catalog#10061-H08B1; NP_003255.2; Met1-Arg587) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-1694