Learn More
Sino Biological Anti-CDC42 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10062T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CDC42 (rh CDC42; Catalog#10062-H09E; P60953-2; Met1-Cys188). CDC42 specific IgG was purified by Human CDC42 affinity chromatography.
Catalog No. 50-161-1705