Learn More
Sino Biological Recombinant Human BACE1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10064H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (amino acid residue Met 1-Thr 457) of human BACE1 (NP_036236.1) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3923