Learn More
Sino Biological Recombinant Anti-BACE1 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10064R027F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human BACE1 / ASP2 (rh BACE1 / ASP2; Catalog#10064-H08H; NP_036236.1; Met 1-Thr 457) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-1731