Learn More
Sino Biological Anti-IRF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100646T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse IRF6 / IRF-6, and purified by antigen affinity chromatography.
Catalog No. 50-161-1720