Learn More
Sino Biological Anti-ALK-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10066RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ALK-1 / ACVRL1 (rh ALK-1 / ACVRL1; Catalog#10066-H08H; NP_000011.2; Met1-Gln118). ALK-1 / ACVRL1 specific IgG was purified by Human ALK-1 / ACVRL1 affinity chromatography.
Catalog No. 50-161-1778